Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Methyl Violet 2B

Catalog No. p-7081952
Click to view available options
Quantity:
50 g
250 g

CAS: 8004-87-3 | C24H27N3·HCl

Methyl violet 2B is a biological stain that is commonly used at a pH between 0 and 1.6. It is certified for use in Flemming triple stain with iodine for chromosomes; Newton?s crystal violet-iodine technique for chromatin and nucleoli; Lieb?s amyloid stain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl violet 2B is a biological stain that is commonly used at a pH between 0 and 1.6. It is certified for use in Flemming triple stain with iodine for chromosomes; Newton′s crystal violet-iodine technique for chromatin and nucleoli; Lieb′s amyloid stain.

Solubility
Soluble in water.

Notes
Transition interval: pH 0.0 (yellow) to pH 1.6 (blue)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8004-87-3
Molecular Formula C24H27N3·HCl
MDL Number MFCD00012568
Synonym Basic Violet 1; C.I. 42535

Specifications

Solubility Information Soluble in water.
Formula Weight 393.96
Odor Odorless
Quantity 50 g
Chemical Name or Material Methyl Violet 2B
Hazard Category H302-H319
Hazard Statement GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
RTECSNumber GC8425000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.