Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Methylated Lysine Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA577770
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA577770 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA577770 Supplier Invitrogen™ Supplier No. PA577770
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

0.2-0.5 μg/mL of PA5-77770 was sufficient for detection of the methylated histone by western blot analysis using melanoma cells in TBSt. Detects proteins containing methylated lysine residues.

Methylated lysine is derivative of the amino acid residue lysine where the sidechain ammonium group has been methylated one or more times.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Methylated Lysine
Applications ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and no preservative; pH 7.4
Gene Alias Dimethylysine; Methyl lysine; Trimethyl lysine
Host Species Rabbit
Immunogen Methylated KLH Conjugated.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.