Learn More
Invitrogen™ Methylboldenone Polyclonal Antibody

Description
Working Concentration: 0.3125 μg/mL. Sensitivity: 10 ng/mL 17 beta methyltestosterone produces 87% inhibition in a competitive ELISA employing Methylboldenone/Methyltestosterone polyclonal antibody.
Specifications
Specifications
| Antigen | Methylboldenone |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Host Species | Sheep |
| Immunogen | 17B-Methylboldenone(1a)-BTG. |
| Purification Method | Purified |
| Quantity | 1 mg |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.