Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylboronic acid, 97%, Acros Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
13061-96-6
CH5BO2
59.86
MFCD00002105
KTMKRRPZPWUYKK-UHFFFAOYSA-N
methaneboronic acid,methyl boronic acid,dihydroxymethylborane,boronic acid, methyl,unii-8z1me41sch,methane boronic acid,8z1me41sch,methylboronicacid,methyboronic acid,methyl-boronic acid
139377
methylboronic acid
B(C)(O)O
13061-96-6
CH5BO2
59.86
MFCD00002105
KTMKRRPZPWUYKK-UHFFFAOYSA-N
methaneboronic acid,methyl boronic acid,dihydroxymethylborane,boronic acid, methyl,unii-8z1me41sch,methane boronic acid,8z1me41sch,methylboronicacid,methyboronic acid,methyl-boronic acid
139377
methylboronic acid
B(C)(O)O
Quantity 1g
Color White
Infrared Spectrum Authentic
Melting Point 88.0°C to 90.0°C
Packaging Glass bottle
Assay Percent Range 97%
Linear Formula CH3B(OH)2
Solubility Information Solubility in water: soluble in water
Formula Weight 59.86
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material Methylboronic acid
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning