Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylcatechol, 0.05%, Reagents

Catalog No. 01334768
Change view
Click to view available options
:
Quantity: 100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-334-768 Quantity: 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-334-768 Supplier Reagents Holdings Llc Supplier No. CM092800100C
Only null left
Add to Cart
Add to Cart

Methylcatechol, 0.05%, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 90-05-1
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Guaiacol
IUPAC Name water
SMILES O

Specifications

Boiling Range 100°C
Color Colorless
Boiling Point 100°C
Packaging Glass Bottle
Quantity 100 mL
Specific Gravity 1.0001
Percent Purity 0.05%
Grade Reagent
Physical Form Liquid
Chemical Name or Material Methylcatechol, 0.05%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.