Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylcobalamin hydrate, 97%

Catalog No. p-7081276
Click to view available options
Quantity:
100 mg
500 mg

CAS: 288315-09-3 | C63H91CoN13O14P- | 1344.405 g/mol

Methylcobalamin hydrate is a coenzyme required in the biosynthesis of methionine. Vitamin (hematopoietic). It acts as a histamine receptor, Alzheimer research.Methylcobalamin is also used in the treatment of peripheral neuropathy, diabetic neuropathy, and as a preliminary treatment for amyotrophic lateral sclerosis. It can be used to prevent or treat pathology arising from a lack of vitamin B12 such as pernicious anemia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methylcobalamin hydrate is a coenzyme required in the biosynthesis of methionine. Vitamin (hematopoietic). It acts as a histamine receptor, Alzheimer research.Methylcobalamin is also used in the treatment of peripheral neuropathy, diabetic neuropathy, and as a preliminary treatment for amyotrophic lateral sclerosis. It can be used to prevent or treat pathology arising from a lack of vitamin B12 such as pernicious anemia.

Solubility
Partially soluble in cold water, hot water.

Notes
Store at -20°C. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 288315-09-3
Molecular Formula C63H91CoN13O14P-
Molecular Weight (g/mol) 1344.405
MDL Number MFCD00149221
InChI Key QXOMMGINPOYKPR-YHKGLSPZSA-L
Synonym methylcobalamin
PubChem CID 134128059
IUPAC Name carbanide;cobalt(2+);[(2S,3R,4S,5R)-5-(5,6-dimethylbenzimidazol-1-yl)-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl] [(2S)-1-[3-[(1S,2S,3S,5Z,7R,10Z,12R,13R,15Z,17R,18R,19S)-2,13,18-tris(2-amino-2-oxoethyl)-7,12,17-tris(3-amino-3-oxopropyl)-3,5,8,8,13,15,18,19-o
SMILES [CH3-].CC1=CC2=C(C=C1C)N(C=N2)C3C(C(C(O3)CO)OP(=O)([O-])OC(C)CNC(=O)CCC4(C(C5C6(C(C(C(=C(C7=NC(=CC8=NC(=C(C4=N5)C)C(C8(C)C)CCC(=O)N)C(C7(C)CC(=O)N)CCC(=O)N)C)[N-]6)CCC(=O)N)(C)CC(=O)N)C)CC(=O)N)C)O.[Co+2]

Specifications

Quantity 100 mg
Solubility Information Partially soluble in cold water,hot water.
Formula Weight 1344.41 (Anhydrous)
Percent Purity 98%
Chemical Name or Material Methylcobalamin hydrate
Hazard Category H350
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 236-535-3
RTECSNumber GG3745000
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.