Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylcyclohexane, 99+%

Catalog No. p-7049948
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3186122 100 g
AA3186136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3186122 Supplier Thermo Scientific Chemicals Supplier No. 03186122
Only null left
Add to Cart

CAS: 108-87-2 | C7H14 | 98.19 g/mol

Methylcyclohexane is used as rubber, paint and varnish, used solvents, oils, such as solvent extraction, which can also be used for organic synthesis, Standards for the calibration of the thermometer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-87-2
Molecular Formula C7H14
Molecular Weight (g/mol) 98.19
MDL Number MFCD00001497
InChI Key UAEPNZWRGJTJPN-UHFFFAOYSA-N
Synonym cyclohexane, methyl, hexahydrotoluene, cyclohexylmethane, toluene hexahydride, hexahydroxytoluene, sextone b, toluene, hexahydro, metylocykloheksan, methyl cyclohexane, methylcyclohexan
PubChem CID 7962
IUPAC Name methylcyclohexane
SMILES CC1CCCCC1

Specifications

Melting Point -126°C
Color Colorless
Density 0.771 g/mL
Boiling Point 101°C
Flash Point -4°C (-25°F)
Refractive Index 1.423
Quantity 100 g
UN Number UN2296
Beilstein 505972
Merck Index 14,6047
Solubility Information Soluble in water 0.1g/L (20°C).
Formula Weight 98.19
Percent Purity ≥99%
Physical Form Liquid
Chemical Name or Material Methylcyclohexane
Show More Show Less
Hazard Category H225-H304-H315-H319-H335-H336
Hazard Statement GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: METHYLCYCLOHEXANE
EINECSNumber 203-624-3
RTECSNumber GV6125000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.