Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylcyclopentane, 95%, ACROS Organics™

Click to view available options
Quantity:
500mL
2.5L
25L
Packaging:
Glass bottle
Metal Drum
96-37-7
C6H12
84.16
MFCD00001382
GDOPTJXRTPNYNR-UHFFFAOYSA-N
cyclopentane, methyl,methyl cyclopentane,methylpentamethylene,unii-5g26cc1ask,methyl-cyclopentane,ccris 6058,hsdb 876,5g26cc1ask,un2298 flammable liquid,3-methylcyclopentyl
7296
methylcyclopentane
CC1CCCC1
96-37-7
C6H12
84.16
MFCD00001382
GDOPTJXRTPNYNR-UHFFFAOYSA-N
cyclopentane, methyl,methyl cyclopentane,methylpentamethylene,unii-5g26cc1ask,methyl-cyclopentane,ccris 6058,hsdb 876,5g26cc1ask,un2298 flammable liquid,3-methylcyclopentyl
7296
methylcyclopentane
CC1CCCC1
Boiling Point 71.0°C
Viscosity 0.505 mPa.s (20°C)
Flash Point −4°C
Melting Point -142.4°C
Quantity 500mL
Density 0.7480g/mL
Color White
Infrared Spectrum Authentic
Refractive Index 1.4077 to 1.4097
Packaging Glass bottle
Specific Gravity 0.748
Assay Percent Range 94% min. (GC)
Linear Formula C5H9CH3
Beilstein 05, 27
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetone,benzene,carbon tetrachloride,,diethyl ether,ethanol,petroleum ether
Formula Weight 84.16
Physical Form Adhering Crystals
Percent Purity 95%
Chemical Name or Material Methylcyclopentane, 95%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
May be fatal if swallowed and enters airways.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 202-503-2

RTECSNumber : GY4640000

TSCA : TSCA