Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-amino-5-chlorobenzoate, 95%, Thermo Scientific™

Catalog No. ac426160010
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle

Chemical Identifiers

CAS 5202-89-1
Molecular Formula C8H8ClNO2
Molecular Weight (g/mol) 185.61
MDL Number MFCD00007837
InChI Key IGHVUURTQGBABT-UHFFFAOYSA-N
Synonym methyl 5-chloroanthranilate, methyl2-amino-5-chlorobenzoate, 5-chloroanthranilic acid methyl ester, benzoic acid, 2-amino-5-chloro-, methyl ester, 2-amino-5-chlorobenzoic acid methyl ester, anthranilic acid, 5-chloro-, methyl ester, 2-amino-5-chloro-benzoic acid methyl ester, pubchem3346, acmc-1axns, methyl-5-chloroanthranilate
PubChem CID 78878
IUPAC Name methyl 2-amino-5-chlorobenzoate
SMILES COC(=O)C1=C(C=CC(=C1)Cl)N

Specifications

Melting Point 66°C to 68°C
Color Beige-Brown to Cream
Boiling Point 168°C to 170°C (22.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H3(NH2)CO2CH3
Quantity 1 g
Formula Weight 185.61
Percent Purity 95%
Physical Form Crystal Powder or Crystals
Chemical Name or Material Methyl 2-amino-5-chlorobenzoate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
EINECSNumber 225-992-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.