Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-cyclopentanonecarboxylate, 96%

Catalog No. p-3758382
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle

CAS: 10472-24-9 | C7H10O3 | 142.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10472-24-9
Molecular Formula C7H10O3
Molecular Weight (g/mol) 142.15
MDL Number MFCD00001411
InChI Key PZBBESSUKAHBHD-UHFFFAOYSA-N
Synonym methyl 2-oxocyclopentanecarboxylate, methyl 2-cyclopentanonecarboxylate, 2-carbmethoxycyclopentanone, methyl cyclopentanone-2-carboxylate, cyclopentanecarboxylic acid, 2-oxo-, methyl ester, 2-methoxy carbonyl cyclopentanone, 2-methoxycarbonylcyclopentanone, 2-oxocyclopentanecarboxylic acid methyl ester, cyclopentanone-2-carboxylic acid methyl ester, mehyl-2-oxocyclopentane
PubChem CID 66328
IUPAC Name methyl 2-oxocyclopentane-1-carboxylate
SMILES COC(=O)C1CCCC1=O

Specifications

Color Colorless to Yellow
Density 1.0540g/mL
Boiling Point 102°C to 104°C (11.0 mmHg)
Flash Point 77°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.4550 to 1.457
Quantity 5 g
Beilstein 10, 597
Specific Gravity 1.054
Solubility Information Solubility in water: >100g/L. Other solubilities: miscible in ethanol
Formula Weight 142.15
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Methyl 2-cyclopentanonecarboxylate
Show More Show Less
EINECSNumber 233-962-7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.