Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyle3-methyl-2-furoate, 98%, Thermo Scientific™

Catalog No. AC356500010
Click to view available options
Quantity:
1 g
Packaging:
Glass Bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6141-57-7
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.14
MDL Number MFCD00014110
InChI Key AQQYRDKMXXSIMP-UHFFFAOYSA-N
Synonym methyl 3-methyl-2-furoate, methyl 3-methylfuroate, methyl 3-methyl-2-furancarboxylate, furancarboxylic acid, methyl-, methyl ester, 3-methyl-2-furancarboxylic acid methyl ester, 3-methyl-furan-2-carboxylic acid methyl ester, 2-furoic acid, 3-methyl-, methyl ester, acmc-20a9f6, methyl-3-methyl-2-furoate, 2-furancarboxylic acid, 3-methyl-, methyl ester
PubChem CID 350231
IUPAC Name methyl 3-methylfuran-2-carboxylate
SMILES COC(=O)C1=C(C)C=CO1

Specifications

Melting Point 32°C to 36°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Quantity 1 g
Formula Weight 140.14
Percent Purity ≥97.5%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material Methyl 3-methyl-2-furoate, 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.