Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl 2-amino-6,7-dihydro[1,3]thiazolo[5,4-c]pyridine-5(4H)-carboxylate, 97%, Thermo Scientific Chemicals

Catalog No. AC438020050
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 365996-05-0
Molecular Formula C11H17N3O2S
Molecular Weight (g/mol) 255.34
InChI Key BMLHPGOMLGKYIJ-UHFFFAOYSA-N
Synonym tert-butyl 2-amino-6,7-dihydrothiazolo 5,4-c pyridine-5 4h-carboxylate, tert-butyl 2-amino-6,7-dihydro 1,3 thiazolo 5,4-c pyridine-5 4h-carboxylate, 5-boc-2-amino-4,5,6,7-tetrahydrothiazolo 5,4-c pyridine, 2-amino-6,7-dihydro-4h-thiazolo 5,4-c pyridine-5-carboxylic acid tert-butyl ester, tert-butyl 2-amino-4h,5h,6h,7h-1,3 thiazolo 5,4-c pyridine-5-carboxylate, 2-amino-5-boc-6,7-dihydro-4h-thiazolo-5,4,c pyridine, tert-butyl 2-amino-4h,6h,7h-1,3 thiazolo 5,4-c pyridine-5-carboxylate, tert-butyl 2-amino-6,7-dihydro-4h-thiazolo 5,4-c pyridine-5-carboxylate, pubchem17893
PubChem CID 11357283
IUPAC Name tert-butyl 2-amino-6,7-dihydro-4H-[1,3]thiazolo[5,4-c]pyridine-5-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC2=C(C1)SC(=N2)N

Specifications

Melting Point 223.5°C to 224.5°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 5 g
Formula Weight 255.34
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material tert-Butyl 2-amino-6,7-dihydro[1,3]thiazolo[5,4-c]pyridine-5(4H)-carboxylate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.