Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl cyclopropanecarboxylate, 98%

Catalog No. p-3758368
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 2868-37-3 | C5H8O2 | 100.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2868-37-3
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00001280
InChI Key PKAHQJNJPDVTDP-UHFFFAOYSA-N
Synonym methyl cyclopropane carboxylate, cyclopropanecarboxylic acid, methyl ester, methylcyclopropanecarboxylate, cyclopropanecarboxylic acid methyl ester, methyl cyclopropylcarboxylate, unii-5uq53f585u, mcpc, pubchem11835, ksc490a7l, 1-methylcyclopropanecarboxylate
PubChem CID 76122
IUPAC Name methyl cyclopropanecarboxylate
SMILES COC(=O)C1CC1

Specifications

Color Colorless
Density 0.9800g/mL
Boiling Point 119.0°C (764.0 mmHg)
Flash Point 17°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4170 to 1.4190
Linear Formula C3H5CO2CH3
Quantity 25 g
Beilstein 09, I, 3
Specific Gravity 0.98
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohols
Formula Weight 100.12
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methyl cyclopropanecarboxylate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye irritation.
Toxic in contact with skin.
Harmful if swallowed.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove
EINECSNumber 220-690-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.