Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl indole-4-carboxylate, 99%, Acros Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
39830-66-5
C10H9NO2
175.19
MFCD00191222
WEAXQUBYRSEBJD-UHFFFAOYSA-N
methyl indole-4-carboxylate,indole-4-carboxylic acid methyl ester,1h-indole-4-carboxylic acid methyl ester,methyl 4-indolecarboxylate,4-indole-carboxylic acid methyl ester,1h-indole-4-carboxylic acid, methyl ester,4-methoxycarbonylindole,4-methoxycarbonyl-1h-indole,indole-4-carboxylate,pubchem1705
2733668
methyl 1H-indole-4-carboxylate
COC(=O)C1=C2C=CNC2=CC=C1
39830-66-5
C10H9NO2
175.19
MFCD00191222
WEAXQUBYRSEBJD-UHFFFAOYSA-N
methyl indole-4-carboxylate,indole-4-carboxylic acid methyl ester,1h-indole-4-carboxylic acid methyl ester,methyl 4-indolecarboxylate,4-indole-carboxylic acid methyl ester,1h-indole-4-carboxylic acid, methyl ester,4-methoxycarbonylindole,4-methoxycarbonyl-1h-indole,indole-4-carboxylate,pubchem1705
2733668
methyl 1H-indole-4-carboxylate
COC(=O)C1=C2C=CNC2=CC=C1
Packaging Glass bottle
Quantity 1g
Color Green to White
Melting Point 67.0°C to 71.0°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Solubility Information Solubility in water: insoluble
Formula Weight 175.19
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material Methyl indole-4-carboxylate
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.