Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylcyclohexylamine, 99%

Catalog No. AC408250025
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Glass bottle
Plastic drum

CAS: 98-94-2 | C8H17N | 127.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-94-2
Molecular Formula C8H17N
Molecular Weight (g/mol) 127.23
MDL Number MFCD00003844
InChI Key SVYKKECYCPFKGB-UHFFFAOYSA-N
Synonym n,n-dimethylcyclohexylamine, cyclohexyldimethylamine, dimethylcyclohexylamine, n-cyclohexyldimethylamine, polycat 8, cyclohexanamine, n,n-dimethyl, dimethylamino cyclohexane, n,n-dimethylaminocyclohexane, n,n-dimethyl-n-cyclohexylamine, cyclohexylamine, n,n-dimethyl
PubChem CID 7415
ChEBI CHEBI:59022
IUPAC Name N,N-dimethylcyclohexanamine
SMILES CN(C)C1CCCCC1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -60.0°C
Density 0.8400g/mL
Boiling Point 160.0°C
Flash Point 37°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic drum
Refractive Index 1.4525 to 1.4545
Quantity 2.5 kg
Specific Gravity 0.84
Solubility Information Solubility in water: 13g/L (20°C)
Viscosity 1.16 mPa.s (25°C)
Formula Weight 127.23
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N, N-Dimethylcyclohexylamine, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.
Flammable liquid and vapor.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF SWALLOWED
EINECSNumber 202-715-5
RTECSNumber GX1198000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.