Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylene Blue, 0.1% (w/v), Ricca Chemical
SDP

Catalog No. p-2916728
Click to view available options
Quantity:
120 mL
500 mL
1 L
Packaging:
Amber Glass Bottle

Specifications

Chemical Name or Material Methylene Blue
CAS 7732-18-5, 7220-79-3
Percent Purity 0.1% W/V
Grade Laboratory
Concentration or Composition (by Analyte or Components) 0.1% (w/v) Aqueous Solution
Packaging Amber Glass Bottle
Color Blue
Quantity 120 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.