Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methylene Blue, 0.1% (w/v), Ricca Chemical

Click to view available options
Quantity:
120 mL
500 mL
1 L
Packaging:
Amber Glass Bottle
Specifications
Specifications
Chemical Name or Material | Methylene Blue |
CAS | 7732-18-5, 7220-79-3 |
Percent Purity | 0.1% W/V |
Grade | Laboratory |
Concentration or Composition (by Analyte or Components) | 0.1% (w/v) Aqueous Solution |
Packaging | Amber Glass Bottle |
Color | Blue |
Quantity | 500 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction