Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylene Blue 1% Aqueous, Innovating Science™
SCI_ED

Catalog No. S001058 Shop All Aldon Products
Click to view available options
Quantity:
100 mL
500 mL

Catalog No. S001058

Supplier: Aldon Corporation IS22078

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 61-73-4, 7732-18-5, 532-32-1
Molecular Weight (g/mol) Mixture

Specifications

Percent Purity 1%
Grade Laboratory
Color Dark Blue
Quantity 100 mL
Chemical Name or Material Methylene blue chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.