Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Methylene Blue
SCI_ED

Catalog No. p-4416703 Shop All Fisherbrand Products
Click to view available options
Quantity:
10 g
25 g
Packaging:
Solid
Biological stain

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 7220-79-3, 7732-18-5
Molecular Formula C16H24ClN3O3S
Molecular Weight (g/mol) 373.90
MDL Number MFCD00150008
InChI Key XQAXGZLFSSPBMK-UHFFFAOYSA-M
Synonym Methylene Blue Chloride, Basic Blue 9
PubChem CID 104827
IUPAC Name 3,7-bis(dimethylamino)-5λ⁴-phenothiazin-5-ylium trihydrate chloride
SMILES O.O.O.[Cl-].CN(C)C1=CC2=[S+]C3=CC(=CC=C3N=C2C=C1)N(C)C

Specifications

Assay Percent Range 1% w/v
Linear Formula C16H18ClN3S·3H2O
Formula Weight 373.9
Packaging Solid
Density ∽1.00g/mL at 20°C
Color Green
Quantity 25 g
Chemical Name or Material Methylene Blue
ShelfLife 5 years
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.