Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Iron(II) sulfate heptahydrate, 99+%, ACS reagent
Click to view available options
Quantity:
5 g
100 g
500 g
3 kg
10 kg
Packaging:
Glass Bottle
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 64.0°C |
Color | Blue or Blue-Green or Green |
Physical Form | Crystals |
Quantity | 100 g |
Assay Percent Range | 99+% |
Linear Formula | FeSO4·7H2O |
Fieser | 01,393; 04,237; 05,308 |
Merck Index | 15,4084 |
Solubility Information | Solubility in water: 25.6 g/100mL (20°C). Other solubilities: insoluble in alcohol |
IUPAC Name | λ²-iron(2+) heptahydrate sulfate |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Warning |
Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. Harmful if swallowed. |
Precautionary Statement | GHS P Statement IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse |
Recommended Storage | due to inherent oxidation, ferric ion (Fe3+) content may increase on storage |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction