Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylene Blue Reagent, for Surfactant Analysis, Ricca Chemical
SDP

Catalog No. p-3251946 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle

Catalog No. 4900-16

Supplier: Ricca Chemical Company 490016

Only null left
Add to Cart
Add to Cart

For Analysis of Surfactants

Specifications

Chemical Name or Material Methylene Blue Reagent
CAS 7732-18-5, 10049-21-5, 7664-93-9, 7732-18-5
Grade Laboratory
Packaging Natural Poly Bottle
Color Dark Blue
Quantity 500 mL
Physical Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.