Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Methylene Blue Solution, 1%
SCI_ED

Catalog No. s25431 Shop All Fisherbrand Products
Click to view available options
Quantity:
15 mL
100 mL
500 mL
Biological stain

This product is for educational use - no C of A is available.

Specifications

Chemical Name or Material Methylene Blue Solution, 1%
CAS 7220-79-3, 7732-18-5
Assay Percent Range 1% w/v
Linear Formula C16H18ClN3S·3H2O
Synonym Methylene Blue Chloride, Basic Blue 9
Formula Weight 373.9
Packaging 1%
Density ∽1.00g/mL at 20°C
Color Blue
Quantity 100 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.