Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylene Blue trihydrate

Catalog No. p-7058190
Click to view available options
Quantity:
25 g
100 g
250 g

Sensitive stain for RNA and ribonuclease | CAS: 7220-79-3 | C16H24ClN3O3S | 373.90 g/mol

Methylene Blue trihydrate finds many uses in biology and chemistry. It acts as a GCS inhibitor, biological stain and redox indicator. It inhibits tau filament formation. Further, it is reported to bind to the M2 subtype of muscarinic receptors in rat cardiac myocytes and thus exhibit antimuscarinic effects on IK, ACh and ICa. It also inhibits endothelium-dependent relaxation of isolated blood vessels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methylene Blue trihydrate finds many uses in biology and chemistry. It acts as a GCS inhibitor, biological stain and redox indicator. It inhibits tau filament formation. Further, it is reported to bind to the M2 subtype of muscarinic receptors in rat cardiac myocytes and thus exhibit antimuscarinic effects on IK, ACh and ICa. It also inhibits endothelium-dependent relaxation of isolated blood vessels.

Solubility
Soluble in water, alcohol and chloroform. Insoluble in ether.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7220-79-3
Molecular Formula C16H24ClN3O3S
Molecular Weight (g/mol) 373.90
MDL Number MFCD00150008
InChI Key XQAXGZLFSSPBMK-UHFFFAOYSA-M
Synonym C.I. 52015; Basic Blue 9
PubChem CID 104827
IUPAC Name [7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride;trihydrate
SMILES O.O.O.[Cl-].CN(C)C1=CC2=[S+]C3=CC(=CC=C3N=C2C=C1)N(C)C

Specifications

Beilstein 3599847
Merck Index 14,6060
Solubility Information Soluble in water,alcohol and chloroform; Insoluble in ether.
Formula Weight 373.90 (319.86 Anhydrous)
Odor Odorless
Color Green
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Methylene Blue trihydrate
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 200-515-2
RTECSNumber SP5740000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.