Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylene Violet, Certified Bernthsen, Thermo Scientific™

Catalog No. AC414300050
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2516-05-4
Molecular Formula C14H12N2OS
Molecular Weight (g/mol) 256.32
MDL Number MFCD00005064
InChI Key ALJHHTHBYJROOG-UHFFFAOYSA-N
Synonym C.I. 52041

Specifications

Melting Point 216.0°C
Absorption Ratio 0.99 to 1.06
Beilstein 27, II, 463
SMILES CN(C)C1=CC2=C(C=C1)N=C3C=CC(=O)C=C3S2
Infrared Spectrum Authentic
IUPAC Name 7-(dimethylamino)phenothiazin-3-one
PubChem CID 73024
Lambda Max 579 to 584nm
Formula Weight 256.32
Grade Pure
Packaging Glass bottle
Color Green
Quantity 5 g
Physical Form Powder
Chemical Name or Material Methylene Violet, Bernthsen
Show More Show Less
EINECSNumber 219-733-4
TSCA TSCA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.