Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pentylmagnesium bromide, 2M solution in diethyl ether, AcroSeal™
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Black to Yellow |
| Density | 0.9470g/mL |
| Flash Point | -40°C |
| Packaging | AcroSeal™ Glass Bottle |
| Quantity | 100 mL |
| Specific Gravity | 0.947 |
| Solubility Information | Solubility in water: reacts |
| Formula Weight | 175.35 |
| Physical Form | Solution |
| Chemical Name or Material | Pentylmagnesium bromide |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Extremely flammable liquid and vapour. Causes severe skin burns and eye damage. Harmful if swallowed. May cause drowsiness or dizziness. In contact with water releases flammable gases which may ignite sponta |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF IN EYES: |
| EINECSNumber | 211-747-9 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction