Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylepentadecafluorooctanoate, 95%, Thermo Scientific™

Catalog No. AC367650050
Click to view available options
Quantity:
5 mL
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 376-27-2
Molecular Formula C9H3F15O2
Molecular Weight (g/mol) 428.097
MDL Number MFCD00039244
InChI Key XOCNYZFAMHDXJK-UHFFFAOYSA-N
Synonym methyl perfluorooctanoate, methyl pentadecafluorooctanoate, pentadecafluorooctanoic acid, methyl ester, methyl perfluorocaprylate, octanoic acid, pentadecafluoro-, methyl ester, perfluorooctanoic acid methyl ester, pentadecafluorooctanoic acid methyl ester, octanoic acid, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro-, methyl ester, acmc-1cndk, methylpentadecafluorooctanoate
PubChem CID 67824
IUPAC Name methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctanoate
SMILES COC(=O)C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

Color Colorless
Density 1.6840g/mL
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Refractive Index 1.3030 to 1.3070 (20°C, 589nm)
Linear Formula CF3(CF2)6CO2CH3
Quantity 5 mL
Formula Weight 428.1
Percent Purity ≥94%
Physical Form Liquid
Chemical Name or Material Methyl pentadecafluorooctanoate, 95%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.