Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl picolinate, 98%

Catalog No. p-4228824
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 2459-07-6 | C7H7NO2 | 137.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2459-07-6
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
InChI Key NMMIHXMBOZYNET-UHFFFAOYSA-N
SMILES COC(=O)C1=CC=CC=N1

Specifications

Melting Point 14°C to 16°C
Color White
Density 1.1370g/mL
Boiling Point 95°C (1.0 hPa)
Flash Point 103°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Specific Gravity 1.137
Synonym methyl isonicotinate, isonicotinic acid methyl ester, 4-pyridinecarboxylic acid, methyl ester, 4-picolinic acid methyl ester, 4-methoxycarbonylpyridine, methylisonicotinate, methyl 4-pyridinecarboxylate, 4-carbomethoxypyridine, isonicotinic acid, methyl ester, i-nicotinic acid, methyl ester
IUPAC Name methyl pyridine-4-carboxylate
PubChem CID 227085
Formula Weight 137.14
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Methyl picolinate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.
EINECSNumber 219-545-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.