Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl propiolate, 99%, Acros Organics

Click to view available options
Quantity:
10g
50g
250g
Packaging:
Glass bottle
922-67-8
C4H4O2
84.07
MFCD00008572
IMAKHNTVDGLIRY-UHFFFAOYSA-N
methyl propiolate,methyl propynoate,2-propynoic acid, methyl ester,propiolic acid, methyl ester,methyl acetylenecarboxylate,methyl ethynecarboxylate,propynoic acid, methyl ester,propynoic acid methyl ester,methyl propargylate,acetylenecarboxylic acid methyl ester
13536
methyl prop-2-ynoate
COC(=O)C#C
922-67-8
C4H4O2
84.07
MFCD00008572
IMAKHNTVDGLIRY-UHFFFAOYSA-N
methyl propiolate,methyl propynoate,2-propynoic acid, methyl ester,propiolic acid, methyl ester,methyl acetylenecarboxylate,methyl ethynecarboxylate,propynoic acid, methyl ester,propynoic acid methyl ester,methyl propargylate,acetylenecarboxylic acid methyl ester
13536
methyl prop-2-ynoate
COC(=O)C#C
Refractive Index 1.4070 to 1.409
Flash Point 10°C
Boiling Point 98°C to 101°C
Specific Gravity 0.94
Density 0.9400g/mL
Color Colorless
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 10g
Assay Percent Range 99%
Linear Formula HC∽CCO2CH3
Beilstein 02, I, 208
Solubility Information Solubility in water: insoluble
Formula Weight 84.07
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Methyl propiolate
Show More Show Less

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Highly flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond container and receiving equipment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautio

GHS Signal Word: Danger

EINECSNumber : 213-083-5