Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl propiolate, 99%

Catalog No. AC174042500
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle

CAS: 922-67-8 | C4H4O2 | 84.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 922-67-8
Molecular Formula C4H4O2
Molecular Weight (g/mol) 84.07
MDL Number MFCD00008572
InChI Key IMAKHNTVDGLIRY-UHFFFAOYSA-N
Synonym methyl propiolate, methyl propynoate, 2-propynoic acid, methyl ester, propiolic acid, methyl ester, methyl acetylenecarboxylate, methyl ethynecarboxylate, propynoic acid, methyl ester, propynoic acid methyl ester, methyl propargylate, acetylenecarboxylic acid methyl ester
PubChem CID 13536
IUPAC Name methyl prop-2-ynoate
SMILES COC(=O)C#C

Specifications

Color Colorless
Density 0.9400g/mL
Boiling Point 98°C to 101°C
Flash Point 10°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4070 to 1.409
Linear Formula HC∽CCO2CH3
Quantity 250 g
Beilstein 02, I, 208
Specific Gravity 0.94
Solubility Information Solubility in water: insoluble
Formula Weight 84.07
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl propiolate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond container and receiving equipment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautio
EINECSNumber 213-083-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.