Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl succinyl chloride, 97%

Catalog No. AC108401000
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle

CAS: 1490-25-1 | C5H7ClO3 | 150.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1490-25-1
Molecular Formula C5H7ClO3
Molecular Weight (g/mol) 150.56
MDL Number MFCD00000750
InChI Key SRXOJMOGPYFZKC-UHFFFAOYSA-N
Synonym methyl 4-chloro-4-oxobutyrate, methyl 3-chloroformyl propionate, 3-carbomethoxypropionyl chloride, methyl 4-chloro-4-oxo-butanoate, 3-carbomethoxy propionyl chloride, butanoic acid, 4-chloro-4-oxo-, methyl ester, methoxycarbonylpropionyl chloride, 4-chloro-4-oxo-butanoicacimethylester, succinic acid monomethyl ester chloride, 4-chloro-4-oxobutyric acid methyl ester
PubChem CID 73888
IUPAC Name methyl 4-chloro-4-oxobutanoate
SMILES COC(=O)CCC(=O)Cl

Specifications

Density 1.2320g/mL
Boiling Point 58°C to 65°C (3.0 mmHg)
Flash Point 73°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4402
Linear Formula CH3O2CCH2CH2COCl
Quantity 100 g
Beilstein 02, II, 553
Specific Gravity 1.232
Formula Weight 150.56
Percent Purity 97%
Chemical Name or Material Methyl succinyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 216-077-

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.