Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carboxymethoxylamine HemiHydroChloride, 98%

Catalog No. AC108430025
Click to view available options
Quantity:
2.5 g
10 g
50 g
Packaging:
Glass bottle

Catalog No. AC108430025

Supplier: Thermo Scientific Chemicals 108430025

Only null left
Add to Cart

CAS: 2921-14-4 | C4H11ClN2O6 | 218.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2921-14-4
Molecular Formula C4H11ClN2O6
Molecular Weight (g/mol) 218.59
MDL Number MFCD00012955
InChI Key AXIKLJPWEGNZIG-UHFFFAOYSA-N
Synonym o-carboxymethyl hydroxylamine hemihydrochloride, carboxymethoxylamine hemihydrochloride, aminooxyacetic acid hemihydrochloride, 2-aminooxy acetic acid hydrochloride 2:1, unii-5g5qi2gn7a, acetic acid, aminooxy-, hydrochloride 2:1, aminooxy acetic acid hemihydrochloride, 5g5qi2gn7a, carboxymethoxyamine hemihydrochloride
PubChem CID 2723609
IUPAC Name 2-aminooxyacetic acid;hydrochloride
SMILES [Cl-].NOCC(O)=O.[NH3+]OCC(O)=O

Specifications

Melting Point 156.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (H2NOCH2CO2H)2·HCl
Quantity 2.5 g
Solubility Information Solubility in water: decomposes.
Formula Weight 109.3
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Carboxymethoxylamine hemihydrochloride
Show More Show Less
EINECSNumber 220-862-3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.