Learn More
Methylglyoxal Antibody (9E7), DyLight 650, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP259368C
Catalog No. NBP259368C
Only null left
Add to Cart
Description
Description
Methylglyoxal Monoclonal specifically detects Methylglyoxal in Non-species specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Methylglyoxal
Monoclonal
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
GLO1
Synthetic Methylglyoxal modified Keyhole Limpet Hemocyanin (KLH).
100 μL
Specific for Methylglyoxal modified proteins. Does not detect free Methylglyoxal. Does not cross-react with Acrolein, Hexanoyl Lysine, Malondialdeyhde, 4-Hydroxy-2-hexenal, 4-Hydroxy nonenal, or Crotonaldehyde modified proteins.
Store at 4C in the dark.
IgG2a
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
90000000
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
2-oxopropanal, Pyruvaldehyde
Mouse
Protein G purified
Primary
Human, Non-species specific
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.