Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methylglyoxal Antibody (9E7), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259368F
Description
Methylglyoxal Monoclonal specifically detects Methylglyoxal in Non-species specific samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Methylglyoxal | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
GLO1 | |
Synthetic Methylglyoxal modified Keyhole Limpet Hemocyanin (KLH). | |
100 μL | |
Specific for Methylglyoxal modified proteins. Does not detect free Methylglyoxal. Does not cross-react with Acrolein, Hexanoyl Lysine, Malondialdeyhde, 4-Hydroxy-2-hexenal, 4-Hydroxy nonenal, or Crotonaldehyde modified proteins. | |
Store at 4C in the dark. | |
IgG2a |
Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
90000000 | |
Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
2-oxopropanal, Pyruvaldehyde | |
Mouse | |
Protein G purified | |
Primary | |
Human, Non-species specific | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction