Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methylglyoxal Antibody (9E7), Novus Biologicals™

Mouse Monoclonal Antibody
$622.00
Specifications
Antigen | Methylglyoxal |
---|---|
Clone | 90000000 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 |
Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
Description
Methylglyoxal Monoclonal specifically detects Methylglyoxal in Non-species specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Methylglyoxal | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Cancer | |
2739 | |
Synthetic Methylglyoxal modified Keyhole Limpet Hemocyanin (KLH). | |
Primary | |
Specific for Methylglyoxal modified proteins. Does not detect free Methylglyoxal. Does not cross-react with Acrolein, Hexanoyl Lysine, Malondialdeyhde, 4-Hydroxy-2-hexenal, 4-Hydroxy nonenal, or Crotonaldehyde modified proteins. |
90000000 | |
Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
Monoclonal | |
Purified | |
RUO | |
2-oxopropanal, Pyruvaldehyde | |
GLO1 | |
IgG2a | |
Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title