Learn More
Invitrogen™ Methylglyoxal Monoclonal Antibody (9E7), FITC
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA545810
Description
A 1:1,000 dilution of MA5-45810 was sufficient for detection of Methylglyoxal in 0.5 μg of Methylglyoxal conjugated to BSA by ECL immunoblot analysis using Goat Anti-Mouse IgG:HRP as the secondary Antibody. Specific for Methylglyoxal modified proteins. Does not detect free Methylglyoxal. Does not cross-react with Acrolein, Hexanoyl Lysine, Malondialdeyhde, 4-Hydroxy-2-hexenal, 4-Hydroxy nonenal, or Crotonaldehyde modified proteins.
Methylglyoxal, also called pyruvaldehyde or 2-oxopropanal, is the organic compound with the formula CH3C(O)CHO. Gaseous methylglyoxal has two carbonyl groups, an aldehyde and a ketone but in the presence of water, it exists as hydrates and oligomers. Methylglyoxal is a reduced derivative of pyruvic acid.
Specifications
Methylglyoxal | |
Monoclonal | |
1 mg/mL | |
PBS, 9.09mM sodium bicarbonate with 640.91mM DMSO, 136.36mM ethanolamine and no preservative; pH 7.4 | |
Mouse | |
Protein G | |
RUO | |
Chemical | |
Antibody | |
IgG2a |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
9E7 | |
FITC | |
2 oxo propanal; 2-Oxopropanal; Aldoketomutase; epididymis secretory protein Li 74; GLO1; GLOD1; Glx I; GLYI; glyoxalase domain containing 1; glyoxalase I; HEL-S-74; ketone-aldehyde mutase; lactoyl glutathione lyase; lactoylglutathione lyase; Methylglyoxal; Methylglyoxal (MG); Methylglyoxalase; MG; MG-modified protein; Pyruvaldehyde; S-D-lactoylglutathione methylglyoxal lyase | |
Synthetic Methylglyoxal modified Keyhole Limpet Hemocyanin (KLH). | |
100 μg | |
Primary | |
4°C, store in dark | |
Liquid | |
Chemical |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.