Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylglyoxal Mouse anti-Human, ATTO 594, Clone: 9E7, Abnova™

Catalog No. 89338824 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-338-824 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-338-824 Supplier Abnova Corporation Supplier No. MAB19320
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

Methylglyoxal Monoclonal antibody specifically detects Methylglyoxal in Human samples. It is validated for WB, ELISA, ICC, IF, Flow Cyt.

Specifications

Antigen Methylglyoxal
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Flow Cytometry
Classification Monoclonal
Clone 9E7
Conjugate Atto 594
Host Species Mouse
Immunogen Synthetic Methylglyoxal modified Keyhole Limpet Hemocyanin (KLH)
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage Store at -20°C
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.