Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylglyoxal, MP Biomedicals™

Catalog No. ICN15555880 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
250 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15555880 100 mL
ICN15555825 25 mL
ICN15555883 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15555880 Supplier MP Biomedicals, Inc Supplier No. 0215555880
Add to Cart
Add to Cart

Chemical Identifiers

CAS 78-98-8
Molecular Formula C3H4O2
Molecular Weight (g/mol) 72.06
MDL Number MFCD00006960
InChI Key AIJULSRZWUXGPQ-UHFFFAOYSA-N
Synonym methylglyoxal, pyruvaldehyde, pyruvic aldehyde, acetylformaldehyde, acetylformyl, propanal, 2-oxo, 2-ketopropionaldehyde, propanedione, propanolone, methyl glyoxal
PubChem CID 880
ChEBI CHEBI:17158
IUPAC Name 2-oxopropanal
SMILES CC(=O)C=O

Specifications

Color Colorless
Density 1.17g/mL
Quantity 100 mL
Formula Weight 72.06
Physical Form Liquid
Chemical Name or Material Methylglyoxal, Molecular biology
EINECSNumber 201-164-8
Recommended Storage Store at 4°C, dessicated.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.