Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methyllithium, 1-2M in ether, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™
Click to view available options
Quantity:
100 mL
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Packaged under Argon

Specifications
Specifications
Color | Colorless |
Density | 0.732 g/mL |
Flash Point | -17°C (1°F) |
Odor | Ether-like |
Packaging | ChemSeal™ bottles |
Quantity | 100 mL |
UN Number | UN3394 |
Sensitivity | Air and moisture sensitive |
Formula Weight | 21.97 |
Physical Form | Liquid |
Show More |
Safety and Handling
Hazard Category | H224-H250-H260-H302-H314-H335-H336 |
Hazard Statement | GHS H Statement H224-H250-H260-H252-H314-H318-H302-H336 Extremely flammable liquid and vapor. Catches fire spontaneously if exposed to air. In contact with water releases flammable gases which may ignite spontaneously. Self-heating in large quantities; may catch fire. Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. May cause drowsiness or dizziness. |
Precautionary Statement | P210-P222-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P302+P334-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 4.2; Packaging Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, PYROPHORIC, WATER-REACTIVE |
EINECSNumber | 213-026-4 |
TSCA | Yes |
storageNote1 |
|
Recommended Storage | Ambient temperatures |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction