Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyllithium, 1-2M in ether, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™

Catalog No. p-7052186
Click to view available options
Quantity:
100 mL

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 917-54-4
Molecular Formula CH3Li
Molecular Weight (g/mol) 21.98
MDL Number MFCD00008253
InChI Key DVSDBMFJEQPWNO-UHFFFAOYSA-N
Synonym methyllithium, lithium, methyl, methyl lithium, lithium methanide, meli, lithium methyl, lithium carbanide, lithium methide, methllithium, methyllithum
PubChem CID 2724049
IUPAC Name methyllithium
SMILES [Li]C

Specifications

Color Colorless
Density 0.732 g/mL
Flash Point -17°C (1°F)
Odor Ether-like
Packaging ChemSeal™ bottles
Quantity 100 mL
UN Number UN3394
Sensitivity Air and moisture sensitive
Formula Weight 21.97
Physical Form Liquid
Chemical Name or Material Methyllithium
Show More Show Less
Hazard Category H224-H250-H260-H302-H314-H335-H336
Hazard Statement GHS H Statement
H224-H250-H260-H252-H314-H318-H302-H336
Extremely flammable liquid and vapor.
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gases which may ignite spontaneously.
Self-heating in large quantities; may catch fire.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause drowsiness or dizziness.
Precautionary Statement P210-P222-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P302+P334-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.2; Packaging Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, PYROPHORIC, WATER-REACTIVE
EINECSNumber 213-026-4
TSCA Yes
storageNote1
  • Packaged under Argon
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.