Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methyllithium, 1.6M sol. in diethyl ether (± 5% w/v), AcroSeal™, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Colorless to Yellow |
| Linear Formula | CH3Li |
| Fieser | 01,686; 02,274; 03,202; 05,448; 06,384; 07,242; 08,342; 09,311; 11,340; 12,324; 16,219 |
| Density | 0.7000g/mL |
| Solubility Information | Solubility in water: reacts. |
| IUPAC Name | lithium;carbanide |
| Formula Weight | 21.96 |
| Percent Purity | 4.5 to 5.5% (as MeLi) |
| Packaging | AcroSeal™ Glass bottle |
| Specific Gravity | 0.7 |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement Extremely flammable liquid and vapor. Catches fire spontaneously if exposed to air. In contact with water releases flammable gases which may ignite spontaneously. Causes severe skin burns and eye damage. May cause drowsiness or dizziness. Harmful if swallowed. Reacts violently with water. May form explosive peroxides. Repeated exposure may cause skin dryness or cracking. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Store in a dry place. Store in a closed container. IF ON SKIN: Gently wash with plenty of soap and water. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. |
| EINECSNumber | 213-026-4 |
| TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction