Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylmagnesium bromide, 3M solution in diethyl ether, AcroSeal™

Catalog No. AC183548000
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle

CAS: 75-16-1 | CH3BrMg | 119.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-16-1, 60-29-7
Molecular Formula CH3BrMg
Molecular Weight (g/mol) 119.24
MDL Number MFCD00000041
InChI Key AVFUHBJCUUTGCD-UHFFFAOYSA-M
Synonym methylmagnesium bromide, methyl magnesium bromide, magnesium, bromomethyl, bromo methyl magnesium, methylmagnesium bromide solution, 3.0 m in diethyl ether, unii-22cw9773df, memgbr, methymagnesiumbromide, ch3mgbr, methylmagnesiumbromide, grignard reagent
PubChem CID 6349
SMILES C[Mg]Br

Specifications

Color Colorless to Yellow
Linear Formula CH3MgBr
Concentration 2.85 to 3.20M
Density 1.0350g/mL
Solubility Information Solubility in water: vigorous reaction.
IUPAC Name bromo(methyl)magnesium
Formula Weight 119.26
Packaging AcroSeal™ Glass Bottle
Specific Gravity 1.035
Quantity 800 mL
Flash Point −40°C
Physical Form Solution, may develop some turbidity precipitate
Chemical Name or Material Methylmagnesium bromide, 3M Solution in Diethyl Ether
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage. May cause drowsiness or dizziness. Harmful if swallowed. In contact with water releases flammable gases which may ignite spontaneously. Extremely flammable liquid and vapor. May form explosive peroxides. Reacts violently with water.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Immediately call a POISON CENTER or doctor/physician. Keep away from any possible contact with water, because of violent reaction and possible flash fire.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.