Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylmagnesium bromide, 1M solution in CPME, AcroSeal™

Catalog No. ac446001000
Click to view available options
Quantity:
100 mL
800 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 75-16-1 | CH3BrMg | 119.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-16-1, 5614-37-9
Molecular Formula CH3BrMg
Molecular Weight (g/mol) 119.24
MDL Number MFCD00000041
InChI Key AVFUHBJCUUTGCD-UHFFFAOYSA-M
Synonym methylmagnesium bromide, methyl magnesium bromide, magnesium, bromomethyl, bromo methyl magnesium, methylmagnesium bromide solution, 3.0 m in diethyl ether, unii-22cw9773df, memgbr, methymagnesiumbromide, ch3mgbr, methylmagnesiumbromide, grignard reagent
PubChem CID 6349
IUPAC Name magnesium;carbanide;bromide
SMILES C[Mg]Br

Specifications

Quantity 100 mL
Linear Formula CH3MgBr
Flash Point −1°C
Solubility Information Solubility in water: reacts
Formula Weight 119.24
Chemical Name or Material Methylmagnesium bromide
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapour.
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful to aquatic life with
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomi
EINECSNumber 200-844-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.