Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methylmalonyl Coenzyme A mutase Antibody (OTI2C8), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272652FR
Description
Methylmalonyl Coenzyme A mutase Monoclonal specifically detects Methylmalonyl Coenzyme A mutase in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Methylmalonyl Coenzyme A mutase | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
MMUT | |
Full length recombinant protein of human Methylmalonyl Coenzyme A mutase (NP_000246) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG2a |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI2C8 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
EC 5.4.99.2, MCMmethylmalonyl Coenzyme A mutase, methylmalonyl CoA mutase, Methylmalonyl-CoA isomerase, methylmalonyl-CoA mutase, mitochondrial | |
Mouse | |
Affinity Purified | |
Lipid and Metabolism | |
4594.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction