Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methylparaben, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
100 g
500 g
5 kg
50 kg
Packaging:
Poly Bottle
Specifications
Specifications
| CAS Max % | 1 |
| Boiling Point | 270 to 280°C |
| Packaging | Poly Bottle |
| Quantity | 100 g |
| Grade | Reagent |
| Physical Form | Crystalline Powder or Lumps |
CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction