Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylphosphonic acid, 98%, Thermo Scientific Chemicals

Catalog No. aaa1261903
Click to view available options
Quantity:
1 g
5 g
25 g
100 g

Methylphosphonic acid is used to produce methyl-phosphonic acid dichloride by chlorination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methylphosphonic acid is used to produce methyl-phosphonic acid dichloride by chlorination.

Solubility
Soluble in water and alcohol.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 993-13-5
Molecular Formula CH5O3P
Molecular Weight (g/mol) 96.02
MDL Number MFCD00002137
InChI Key YACKEPLHDIMKIO-UHFFFAOYSA-N
Synonym methanephosphonic acid, phosphonic acid, methyl, methyl phosphonic acid, unii-329w4ym10z, phosphonomethyl group, ester group, phosphonomethyl, acmc-20ake9, d02fxw, methylphosphonic acid
PubChem CID 13818
ChEBI CHEBI:45129
IUPAC Name methylphosphonic acid
SMILES CP(O)(O)=O

Specifications

Melting Point 103°C to 107°C
Quantity 1 g
UN Number UN3261
Beilstein 1739372
Sensitivity Hygroscopic
Solubility Information Soluble in water and alcohol.
Formula Weight 96.03
Percent Purity 98%
Chemical Name or Material Methylphosphonic acid
Hazard Category H302-H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 213-607-2
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.