Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Methylprednisolone Polyclonal Antibody

Sheep Polyclonal Antibody
Supplier: Invitrogen™ PA175286
Description
Working Concentration: 0.625 μg/mL. Sensitivity: 10 ng/mL Methylprednisolone produces 96% inhibition in a competitive ELISA employing Methylprednisolone polyclonal antibody.
Methylprednisolone is a synthetic corticosteroid with anti-inflammatory and immunomodulating properties. Methylprednisolone binds to and activates specific nuclear receptors, resulting in altered gene expression and inhibition of proinflammatory cytokine production. This agent also decreases the number of circulating lymphocytes, induces cell differentiation, and stimulates apoptosis in sensitive tumor cell populations. [National Center for Biotechnology Information. PubChem Compound Database; CID=6741].
Specifications
Methylprednisolone | |
Polyclonal | |
PBS with 0.09% sodium azide; pH 8 | |
Sheep | |
Purified | |
RUO | |
Chemical | |
Antibody | |
IgG |
ELISA | |
Unconjugated | |
C22H30O5; Medrate; Medrol; Medrone; Urbason | |
Methylprednisolone-BTG. | |
1 mg | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction