Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Methylthio)acetaldehyde dimethyl acetal, 97%, Thermo Scientific Chemicals

Catalog No. AAB2266714
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2266714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2266714 Supplier Thermo Scientific Chemicals Supplier No. B2266714

missing translation for 'validMainframeMsg'

CAS: 40015-15-4 | Molecular Formula: C5H12O2S | Molecular Weight: 136.209 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40015-15-4
Molecular Formula C5H12O2S
Molecular Weight (g/mol) 136.209
MDL Number MFCD00009848
InChI Key DVOAQLUDKIFSNB-UHFFFAOYSA-N
Synonym methylthio acetaldehyde dimethyl acetal, 1,1-dimethoxy-2-methylthio ethane, ethane, 1,1-dimethoxy-2-methylthio, 1,1-dimethoxy-2-methylsulfanyl-ethane, ethane,1,1-dimethoxy-2-methylthio, methylmercapto acetaldehyde dimethyl acetal, 1,1-dimethoxy-2-methylsulfanyl ethane, acmc-209j9v, methylthio acetaldehyde dimethylacetal, 2-methylthio acetaldehyde dimethyl acetal
PubChem CID 638113
IUPAC Name 1,1-dimethoxy-2-methylsulfanylethane
SMILES COC(CSC)OC

Specifications

Density 1.022
Boiling Point 170°C to 171°C
Flash Point 53°C (127°F)
Refractive Index 1.4528
Quantity 25 g
UN Number UN1993
Beilstein 773689
Formula Weight 136.21
Percent Purity 97%
Chemical Name or Material (Methylthio)acetaldehyde dimethyl acetal
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H226-H319-H335
Flammable liquid and vapour.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: II
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.