Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Methylthio)acetic acid, 98%

Catalog No. aab2253103
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 2444-37-3 | C3H6O2S | 106.139 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2444-37-3
Molecular Formula C3H6O2S
Molecular Weight (g/mol) 106.139
MDL Number MFCD00075444
InChI Key HGTBAIVLETUVCG-UHFFFAOYSA-N
Synonym methylthio acetic acid, 2-methylthioacetic acid, 2-methylthio acetic acid, acetic acid, methylthio, 2-methylsulfanyl acetic acid, methylsulfenylacetic acid, unii-umv7e1ucux, methylmercaptoacetic acid, umv7e1ucux, acetic acid,2-methylthio
PubChem CID 75551
ChEBI CHEBI:47870
IUPAC Name 2-methylsulfanylacetic acid
SMILES CSCC(=O)O

Specifications

Melting Point 13°C to 14°C
Density 1.224
Boiling Point 99°C to 100°C (4 mmHg)
Flash Point >110°C (230°F)
Odor Stench
Refractive Index 1.495
Quantity 1 g
Beilstein 1740201
Formula Weight 106.15
Percent Purity 98%
Chemical Name or Material (Methylthio)acetic acid
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 219-483-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.