Alfa Aesar™ Methyltin trichloride, 97%

Manufacturer: Alfa Aesar™ 7115606

Quantity 5g

 View more versions of this product

Catalog No. AA7115606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 993-16-8
    Molecular Formula CH3SnCl3
    Formula Weight 240.11
    MDL Number MFCD00000514
    UN Number UN2923
    Synonym Trichloromethylstannane
    Chemical Name or Material Methyltin trichloride

    Specifications

    Formula Weight 240.11
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Crystalline
    Molecular Formula CH3SnCl3
    MDL Number MFCD00000514
    Health Hazard 1 Danger
    Sensitivity Moisture Sensitive
    TSCA TSCA
    CAS 993-16-8
    Synonym Trichloromethylstannane
    Health Hazard 2 GHS H Statement
    H228-H300-H310-H330-H314-H318
    Flammable solid.
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Melting Point 43°C
    Quantity 5g
    UN Number UN2923
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
    Chemical Name or Material Methyltin trichloride
    Assay 97%

    GHS H Statement
    H228-H300-H310-H330-H314-H318
    Flammable solid.
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P210-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit