Learn More
Methyltin trichloride, 97%
Description
A chemical used in the synthesis of metallasilesesquiloxanes. Also used in the preparation of organotin(IV) complexes showing antibacterial activities.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
            Specifications
Specifications
| Melting Point | 43°C | 
| Quantity | 1 g | 
| UN Number | UN2923 | 
| Sensitivity | Moisture sensitive | 
| Solubility Information | Soluble in Chloroform,Methanol. | 
| Formula Weight | 240.11 | 
| Percent Purity | 97% | 
| Physical Form | Crystalline | 
| Chemical Name or Material | Methyltin trichloride | 
Safety and Handling
| Hazard Category | H228-H302-H314-H335 | 
| Hazard Statement | GHS H Statement H228-H300-H310-H330-H314-H318 Flammable solid. Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage.  | 
                                
| Precautionary Statement | P210-P240-P241-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c | 
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S. | 
| EINECSNumber | 213-608-8 | 
| RTECSNumber | WH8585500 | 
| TSCA | Yes | 
| Recommended Storage | Ambient temperatures | 
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.